Learn More
Gilson Company Inc TEST SCREEN TRAY 5/8"
Supplier:  Gilson Company Inc TSA100 5/8
Encompass_Preferred
5/8 inch (16.0mm) full ASTM Testing Screen Trays feature welded and bolted heavy gauge painted steel construction and are fitted with ASTM E11 Compliance Grade wire cloth.Features:Meets ASTM E11 wire cloth specificationsRugged steel Screen Trays are designed for use in Gilson Testing Screen and Test-Master® modelsIncluded Items:5/8” (16.0mm) Testing Screen TraySerial NumberCompliance Grade ASTM E11 Wire Cloth
Catalog No. NC2224888