Learn More
Fritsch Milling And Sizing Inc Test Sieve 200/50 mm Mesh = 0.071 mm Iso 3310-1
SDP
Supplier:  Fritsch Milling And Sizing Inc 30.5800.03
Encompass
test sieve 200/50 mm mesh = 0.071 mm ISO 3310-1
Catalog No. 50-153-4149