Learn More
IBL America Testosterone (Mouse/Rat) ELISA
Supplier:  IBL America IB79174
Encompass_Preferred
Enzyme immunoassay (ELISA) for the determination of Testosterone in rat and mouse serum or plasma. For research use only, not for use in diagnostic procedures.An unknown amount of testosterone present inthe sample and a defined amount of testosterone conjugated to horseradish peroxidase compete for thebinding sites of testosterone antiserum coated to the wells of a microplate. After one-hour incubation on ashaker the microplate is washed four times. After addition of the substrate solution the concentration oftestosterone is inversely proportional to the optical density measured.
Catalog No. 50-153-8189