Learn More
Medchemexpress LLC Tetanus toxin (830-843) TFA | 119260-99-0 | 96.1% | 1725.86 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1754A10MG
Encompass_Preferred
Tetanus toxin (830-843) TFA is a powerful neurotoxin that reaches the cytoplasm of spinal inhibitory interneurons by retroaxonal transport and transcytosis, blocking their ability to release neurotransmitters.
  • Powerful neurotoxin
  • Blocks ability to release neurotransmitters
  • Reaches cytoplasm of spinal inhibitory interneurons by retroaxonal transport and transcytosis
  • Peptide epitope
  • Inhibitor
Catalog No. 50-004-23880