Learn More
Takara Bio TETR MONOCLONAL ANTIBODY 40UG
SDP
Supplier:  Takara Bio 631131
Encompass
Monoclonal Tet antibody to detect Tet-On & Tet-Off Advanced. Antibody for the detection of Tet-On Advanced & Tet-Off Advanced tetracycline transactivators. It also detects the bacterial Tet repressor protein TetR.
Catalog No. NC9960737