Learn More
Strem, An Ascensus Company CAS# 1461-25-2. 50g. Tetra-n-butyltin, min. 94%. MFCD00009422
SDP
Supplier:  Strem, An Ascensus Company 93504950G
Encompass
CAS# 1461-25-2. 50g. Tetra-n-butyltin, min. 94%. MFCD00009422. Molecular Weight: 347.16. Molecular Formula: Sn(C4H9)4. Color/form: colorless liq. Strem# 93-5049. http://www.strem.com/catalog/v/93-5049/
Catalog No. 50-901-15598