Learn More
Enzo Life Sciences Tetra-ubiquitin (K48-linked) (25 µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2660025
Encompass_Preferred
Synonyms: Ub4. MW: ~34kDa. Source: Produced in E. coli. Formulation: Liquid, containing 20mM Tris-HCl, pH 7.5, 0.15M NaCl, 1mM EDTA.
Catalog No. 50-210-0571