Learn More
TARGETMOL CHEMICALS INC TETRABENAZINE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T131345MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (+)-Tetrabenazine ((3R11bR)-Tetrabenazine) is a reversible vesicular monoamine transporter 2 (VMAT-2) inhibitor inhibits transport by VMAT2 with 10-fold greater potency than transport by VMAT1. purity: 98%
Catalog No. 50-258-1377