Learn More
Medchemexpress LLC Tetrabenazine-d6 | 1392826-25-3 | 98.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0590S5MG
Encompass_Preferred
Tetrabenazine-d6 (Deutetrabenazine) is a deuterium-labeled Tetrabenazine, which is the first deuterium approved worldwide for the research of Huntington's disease, or other hyperkinetic movement disorders.
  • Extend the half-life in vivo.
  • Improve oral bioavailability.
  • Improve metabolic characteristics.
  • Improve drug safety.
  • Preserve the therapeutic properties.
Catalog No. 50-004-05883