Learn More
Medchemexpress LLC Tetrabenazine-d6 | 1392826-25-3 | ≥99.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0590S
Encompass_Preferred
Tetrabenazine-d6 (Deutetrabenazine) is a deuterium-labeled Tetrabenazine, the first deuterium approved worldwide for the research of Huntington's disease or other hyperkinetic movement disorders.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Extends the half-life in vivo.
  • Improves oral bioavailability.
  • Improves metabolic characteristics, potentially reducing toxic or reactive metabolites.
  • Improves drug safety by reducing or eliminating adverse side effects.
  • Preserves therapeutic properties, retaining similar biochemical potency and selectivity to hydrogen analogs.
Catalog No. 50-004-50440