Learn More
Abcam Tetrabenazine, VMAT2 inhibitor, 5MG
Supplier:  Abcam AB1461665MG
Encompass_Preferred
MW 317.42 g/mol. Potent, reversible VMAT2 inhibitor (IC₅₀ = 3.2 nM, Ki = 100 nM). Depletes monoamines stores and decreases uptake into synaptic vesicles. Shows antihyperkinetic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4901