Learn More
Medchemexpress LLC Tetrabutylammonium 50g
SDP
Supplier:  Medchemexpress LLC HYW09979250G
Encompass_Preferred
Tetrabutylammonium triiodideIt is an organic compound and belongs to the category of quaternary ammonium salts It is a strong oxidizing agent and iodinating reagent often used in organic chemical reactions especially the conversion of alkenes and alkynes to the corresponding diiodide or tetraiodide compounds Tetrabutylammonium triiodideThere are various applications in synthetic organic chemistry especially in the synthesis of natural products pharmaceuticals and specialty chemicals
Catalog No. 50-002-04862