Learn More
Medchemexpress LLC Tetrabutylammonium bromide | 1643-19-2 | ≥98.0% | 500 G
SDP
Supplier:  Medchemexpress LLC HYD0172500G
Encompass_Preferred
Tetrabutylammonium bromide is an organic ammonium compound primarily used in catalytic reactions and separation/purification processes. It demonstrates significant catalytic effects in certain organic synthesis reactions and acts as a surfactant, stabilizer, and antibacterial agent. In laboratory settings, it is utilized as an ion exchanger and solvent extractant for life science-related research.
  • Significant catalytic effect in organic synthesis reactions
  • Functions as a surfactant and stabilizer
  • Acts as an antibacterial agent
  • Used as an ion exchanger and solvent extractant
  • Biochemical reagent for life science research
Catalog No. 50-004-10016