Learn More
Matrix Scientific Tetrabutylammonium nitrate, 1941-27-1, MFCD00043202, 25g
Supplier: Matrix Scientific 09979225G
Molecular Formula C16H36N2O3, Purity 95+%, Molecular Weight 304.48, Melting Point 116-118°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.