Learn More
Medchemexpress LLC Tetrabutylammonium (tetrahydroborate) | 33725-74-5 | 98% | 257.31 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0086355G
Encompass_Preferred
Tetrabutylammonium tetrahydroborate is an organic compound commonly used as a reducing agent and catalyst. It acts as a reducing agent in chemical reactions, promoting the reduction of organic compounds such as esters, aldehydes, and ketones. It is also widely used in organic synthesis and chemical analysis, with important applications in various production processes.
  • Acts as a reducing agent and catalyst
  • Promotes reduction of organic compounds like esters, aldehydes, and ketones
  • Widely used in organic synthesis
  • Utilized in chemical analysis
Catalog No. 50-003-66684