Learn More
Medchemexpress LLC Tetrabutylammonium trifluoromethanesulfonate | 35895-70-6 | >98.0% | 391.53 g/mol | C17H36F3NO3S | 100 G
SDP
Supplier:  Medchemexpress LLC HYW042557100G
Encompass_Preferred
Tetrabutylammonium trifluoromethanesulfonate is an organic ammonium salt used as a phase-transfer reagent and general-purpose reagent in organic synthesis. It is typically supplied with high purity suitable for research applications.
  • High purity (≥98.0%).
  • Used as a phase-transfer reagent and catalyst in organic reactions.
  • Soluble in common organic solvents.
  • Available in multiple pack sizes, including 100 g.
  • Useful for facilitating nucleophilic substitutions and other synthetic transformations.
Catalog No. 50-002-10231