Learn More
Medchemexpress LLC Tetrachloroaurate II 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW094511
Encompass_Preferred
Tetrachloroaurate III sodium dihydrate (Gold chloride sodium dihydrate) is utilized as catalyst in reactions like nucleophilic addition to multiple bonds nucleophilic substitution of propargylic alcohols and nonsymmetrical etherization[1][2]
Catalog No. 50-002-04645