Learn More
Medchemexpress LLC Tetrachloroauric acid | 16903-35-8 | 97.0% | 339.79 g/mol | HAuCl4 | 1 G

Supplier: Medchemexpress LLC HYW0350911G
Tetrachloroauric acid (HAuCl4) is an inorganic gold(III) salt used as a laboratory reagent and gold source for plating and nanoparticle synthesis. It is supplied in small pack sizes for research applications and should be handled with appropriate corrosive-material precautions and protective measures.
- Molecular weight 339.79 g/mol
- Chemical formula HAuCl4
- Suitable for preparation of gold nanoparticles and gold plating
- Available in small laboratory pack sizes including 1 g
- Comes with analytical data/COA to verify assay and quality
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.