Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrachloroauric acid | 16903-35-8 | MFCD00011322 | 97.0% | 339.79 g/mol | HAuCl4 | 500 MG
SDP

Supplier:  Medchemexpress LLC HYW035091500MG

Encompass_Preferred

Tetrachloroauric acid (HAuCl4) is an inorganic gold(III) chloride used as a source of Au(III) for laboratory applications including gold plating and the preparation of gold nanoparticles. It is supplied as a yellow-orange solid with typical purity around 97% and requires low-temperature, inert storage to maintain stability. Documentation such as a certificate of analysis and safety data sheet is provided.

  • Source of Au(III) for plating and nanoparticle synthesis.
  • Suitable for reagent-grade laboratory use.
  • Yellow-orange solid with high purity.
  • Store at -20°C under nitrogen; in solvent stability varies.
  • Supplied with COA, SDS, and handling documentation.
  • Available in multiple laboratory pack sizes.

Catalog No. 50-003-01546


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.