Learn More
Cayman Chemical TetrachlorohydroquInon 5g
SDP
Supplier:  Cayman Chemical 232315 G
Encompass_Preferred
A metabolite of pentachlorophenol; cytotoxic to RTL-W1 rainbow trout liver cells (EC50 = 1.55 μM in a neutral red assay); increases production of ROS, inhibits apoptosis, and induces loss of the mitochondrial membrane potential and necrosis in splenocytes; induces GSH depletion in mouse liver
Catalog No. 50-298-7212