Learn More
Medchemexpress LLC Tetracycline | 60-54-8 | 98.0% | C22H24N2O8 | 10 G
SDP
Supplier:  Medchemexpress LLC HYA010710G
Encompass_Preferred
Tetracycline is a broad-spectrum, orally active antibiotic. It is effective against a wide range of bacteria, including gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae, making it suitable for infection research.
  • Effective against gram-positive and gram-negative bacteria
  • Active against chlamydiae, mycoplasmas, and rickettsiae
  • Inhibits L-amyloid aggregates formation and disassembles pre-formed fibrils
  • Can be used in animal modeling for genetically engineered disease models
Catalog No. 50-004-33566