Learn More
Alkali Scientific Tetracycline Hydrochloride, 1 Kilogram
SDP
Supplier:  Alkali Scientific TET004CP
Encompass_Preferred
Tetracycline Hydrochloride is a broad-spectrum antibiotic used to treat a variety of bacterial infections. The 1 kg powder form is used for preparing bulk solutions for pharmaceutical, veterinary, and laboratory use. Tetracycline works by inhibiting bacterial protein synthesis, specifically by binding to the 30S ribosomal subunit, preventing the attachment of aminoacyl-tRNA. This action halts the growth of bacterial cells and prevents further proliferation. It is effective against both Gram-positive and Gram-negative bacteria and is commonly used in microbiology labs for bacterial culture studies and resistance testing.
Catalog No. 50-006-61387