Learn More
Medchemexpress LLC Tetracycline (hydrochloride) | 64-75-5 | 98.66% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB04745G
Encompass_Preferred
Tetracycline hydrochloride is a broad-spectrum antibiotic with oral activity. It is effective against a wide range of bacteria, including gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae, making it suitable for research related to infections.
  • Broad-spectrum antibiotic
  • Oral activity
  • Active against gram-positive and gram-negative bacteria
  • Inhibits L-amyloid aggregates formation
  • Can disassembles pre-formed fibrils
  • Suitable for animal modeling in genetically engineered disease models
Catalog No. 50-004-05262