Learn More
Medchemexpress LLC Tetradecaethylene glycol | 1189112-05-7 | 98.0% | 100 MG

Supplier: Medchemexpress LLC HY120430100MG
HO-PEG14-OH is a polyethylene glycol (PEG) linker with terminal hydroxyl groups used as a spacer in the synthesis of PROTACs and other bioconjugates.
- Provides a 14-unit PEG spacer with terminal hydroxyl groups.
- High purity, 98.0%.
- Molecular weight approximately 634.75 g/mol.
- Soluble in DMSO at ≥ 100 mg/mL.
- Colorless to light yellow liquid; density 1.116 g/cm3.
- Storage: -20°C (pure) for long term; -80°C in solvent for extended storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.