Learn More
Medchemexpress LLC Tetradecyltrimethylammonium bromide | 1119-97-7 | 98.0% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYD0839
Encompass_Preferred
Tetradecyltrimethylammonium bromide (TTAB; TTABr; MiTMAB) is a cationic surfactant with an asymmetrical structure, functioning as an organic building block. This laboratory chemical is primarily intended for research use.
  • White to off-white solid appearance
  • 98.0% purity (NMR)
  • Melting point 245-250 °C
  • Relative density 1.1328 g/cm³
Catalog No. 50-004-13937