Learn More
Medchemexpress LLC Tetradifon | 116-29-0 | 99.8% | 25 MG

Supplier: Medchemexpress LLC HY11972525MG
Tetradifon (CAS 116-29-0) is an organochlorine insecticide and acaricide used as a research-grade analytical standard and reagent. It has molecular formula C12H6Cl4O2S and a molecular weight of 356.05 g/mol. Offered as solid vials and solution aliquots for laboratory research and analytical applications.
- Organochlorine insecticide and acaricide.
- Cas number 116-29-0 and molecular formula C12H6Cl4O2S.
- High purity (reported 99.78%).
- Available in solid vials (25 mg-1 g) and 10 mM solutions in DMSO.
- Powder stable at -20°C for up to 3 years; in solution store at -80°C for extended stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.