Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetraethylammonium trifluoromethanesulfonate | 35895-69-3 | MFCD00042586 | ≥98.0% | 279.32 g/mol | C9H20F3NO3S | 10 G
SDP

Supplier:  Medchemexpress LLC HYW12767810G

Encompass_Preferred

Tetraethylammonium trifluoromethanesulfonate is the triflate salt of tetraethylammonium used as a reagent and supporting electrolyte in organic synthesis and electrochemical research. It is a quaternary ammonium salt with a well-defined molecular formula and molecular weight and is supplied for laboratory use with recommended sealed, moisture-free storage.

  • Quaternary ammonium triflate reagent suitable for research use.
  • Molecular formula C9H20F3NO3S; molecular weight 279.32 g/mol.
  • Typical physical form: solid, crystalline.
  • Storage: 4°C, sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
  • CAS number 35895-69-3.

Catalog No. 50-002-82779


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.