Learn More
Medchemexpress LLC Tetraethylene glycol monotosylate | 77544-60-6 | MFCD22056303 | ≥95.0% | 348.41 g·mol⁻¹ | C15H24O7S | 25 G
SDP
Supplier:  Medchemexpress LLC HY4154125G
Encompass_Preferred
Tetraethylene glycol monotosylate is a cleavable linker reagent used in bioconjugation chemistry for preparing antibody-drug conjugates and as a linker in PROTAC constructs. It is supplied as a laboratory-grade chemical suitable for synthetic applications.
  • Cleavable acylhydrazone-based ADC linker
  • Also used as a PROTAC linker in targeted protein degradation
  • Molecular formula C15H24O7S, molecular weight 348.41 g·mol⁻¹
  • Typical purity ≥95%
  • Available in laboratory pack sizes including 25 G
Catalog No. 50-002-03361