Learn More
Medchemexpress LLC Tetraglyme | 143-24-8 | 99.93% | 222.28 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW013522500G
Encompass_Preferred
Me-PEG4-Me is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Is a peg-based protac linker
  • Useful in the synthesis of protacs
Catalog No. 50-004-54233