Learn More
Medchemexpress LLC Tetraheptylammonium iodide | 3535-83-9 | MFCD00041984 | ≥99.0% | 537.69 g/mol | C28H60IN | 10 G

Supplier: Medchemexpress LLC HYW12766110G
Tetraheptylammonium iodide is a quaternary ammonium salt used as a reagent in biochemical research and organic synthesis (CAS 3535-83-9). It is supplied as a solid (white to light-yellow) with molecular formula C28H60IN and molecular weight 537.69 g/mol.
- Quaternary ammonium reagent suitable for biochemical assays and synthesis.
- High purity reported (assay ≥99.0%) for reliable analytical performance.
- Solid, white to light-yellow form for straightforward handling and storage.
- Available in small research sizes for lab-scale use (5 g, 10 g, 25 g).
- Recommended storage at 4°C, sealed; in solution store at -80°C (6 months) or -20°C (1 month).
- Compatible with common organic solvents and assay conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.