Learn More
Medchemexpress LLC HY-N0893 5mg Medchemexpress, Tetrahydrocurcumin CAS:36062-04-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN08935MG
Encompass_Preferred
Medchemexpress, HY-N0893 5mg Tetrahydrocurcumin CAS:36062-04-1 Tetrahydrocurcumin is a Curcuminoid found in turmeric (Curcuma longa) that is produced by the reduction of Curcumin. Tetrahydrocurcumin inhibit CYP2C9 and CYP3A4 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3779