Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran ACS reagent, >=99.0%, contains 250 ppm BHT as inhibitor | 109-99-9 | MFCD00005356 | 4L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 6767644L
Encompass
Tetrahydrofuran ACS reagent, >=99.0%, contains 250 ppm BHT as inhibitor | Purity: >=99.0% | Mol Wt: 72.11 | 109-99-9 | MFCD00005356 | 4L
Catalog No. 50-188-3394