Learn More
Cambridge Isotope Laboratories Tetrahydrofuran-d8 (d, 99.5%), 1693-74-9, MFCD00044238, 10X0.75 mL
Supplier:  Cambridge Isotope Laboratories DLM3610X0.75
Encompass_Preferred
Chemical Formula: C4D8O, MW: 80.16, 99.5%, Synonyms: THF, Deuterated tetrahydrofuran, Octadeuterotetrahydrofuran, THF-d8, 1,4-epoxybutane.
Catalog No. NC9605471