Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran-d8 99.5 at1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1843141G
Encompass
Tetrahydrofuran-d8 may be used as a solvent in the synthesis of deuteriotris[bis(trimethylsilyl)amido]thorium and deuteriotris[bis(trimethylsiiyl)amido]uranium.
Catalog No. 50-289-9198