Learn More
Medchemexpress LLC Tetrahydromagnolol 10mg | 20601-85-8 | 270.37 | C18H22O2 | 10 MG

Supplier: Medchemexpress LLC HY11663710MG
Tetrahydromagnolol is a research-grade small molecule (CAS 20601-85-8) described as a metabolite of magnolol and reported to act as a CB2 receptor agonist and GPR55 antagonist. Supplied as a 10 mg solid for laboratory research use only, it offers high purity and documented solubility and storage recommendations for both in vitro and in vivo studies.
- High purity: 99.89% by HPLC.
- Molecular formula C18H22O2; molecular weight 270.37.
- Soluble in DMSO (100 mg/mL) with established in vivo formulation protocols for administration.
- Recommended storage: solid at 4°C protected from light; in solvent store at -80°C for long term.
- Intended for research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.