Learn More
Medchemexpress LLC Tetrahydrothiophen-3-one | 1003-04-9 | 99.7% | 102.15 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW010594100MG
Encompass_Preferred
Tetrahydrothiophen-3-one is an endogenous metabolite and a pharmaceutical intermediate used to synthesize thiazole compounds. It is found in natural food and has good safety.
  • Endogenous metabolite
  • Pharmaceutical intermediate for synthesizing thiazole compounds
  • Exists in natural food with good safety
  • Appearance: Liquid
  • Color: Colorless to light yellow
Catalog No. 50-004-52898