Learn More
Medchemexpress LLC Tetrahydrothiophen-3-one | 1003-04-9 | 99.7% | 102.15 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW010594500MG
Encompass_Preferred
Tetrahydrothiophen-3-one is an endogenous metabolite and a pharmaceutical intermediate used to synthesize thiazole compounds. It is found in natural food and is considered to have good safety.
  • Endogenous metabolite.
  • Pharmaceutical intermediate that can be used to synthesize thiazole compounds.
  • Exists in natural food with good safety.
Catalog No. 50-003-66530