Learn More
Cayman Chemical TetrahydrourIdIn 10mg
SDP
Supplier:  Cayman Chemical 1640210 MG
Encompass_Preferred
A cytidine deaminase inhibitor (Kis = 54 and 240 nM for the human and E. coli enzymes, respectively); inhibits the metabolism of decitabine and enhances decitabine-induced inhibition of tumor growth in a B16 murine melanoma model
Catalog No. 50-297-1996