Learn More
Medchemexpress LLC Tetrahydrouridine | 18771-50-1 | C9H16N2O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15345A50MG
Encompass_Preferred
Tetrahydrouridine dihydrate is a potent inhibitor of cytidine deaminase (CDA), which competitively blocks the enzyme's active site more effectively than intrinsic cytidine. It is for research use only. Tetrahydrouridine (THU) is a specific inhibitor of cytidine deaminase (CDA) that can suppress deamination in the catabolism of cytotoxic deoxycytidine analogues.
  • Competitively blocks the enzyme's active site more effectively than intrinsic cytidine
  • Can sensitize some pancreatic and lung carcinoma cells to gemcitabine-induced cell death
  • Inhibits S-phase without inducing apoptosis
Catalog No. 50-003-72919