Learn More
Strem, An Ascensus Company CAS 13675-18-8. 25g. Tetrahydroxydiboron min. 95%. MFCD05663888
SDP
Supplier:  Strem, An Ascensus Company 05101525G
Encompass
CAS 13675-18-8. 25g. Tetrahydroxydiboron min. 95%. MFCD05663888. Molecular Weight 89.65. Molecular Formula (OH)2BB(OH)2. Color/form white solid Strem 05-1015. http//www.strem.com/catalog/v/05-1015/
Catalog No. 50-211-0683