Learn More
Medchemexpress LLC Tetrahydrozoline | 84-22-0 | 99.32% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0556
Encompass_Preferred
Tetrahydrozoline (Tetryzoline), a derivative of imidazoline, is an α-adrenergic agonist that causes vasoconstriction. It is widely used for the research of nasal congestion and conjunctival congestion. For research use only.
  • Causes vasoconstriction
  • Used for research of nasal congestion
  • Used for research of conjunctival congestion
  • α-Adrenergic agonist
Catalog No. 50-004-50956