Learn More
Medchemexpress LLC Tetrahydrozoline-d4 hydrochloride | 1246814-66-3 | 99.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0556AS1MG
Encompass_Preferred
Tetrahydrozoline-d4 (hydrochloride) is a deuterium-labeled version of Tetrahydrozoline hydrochloride. Tetrahydrozoline hydrochloride (Tetryzoline hydrochloride) is an imidazoline derivative and an α-adrenergic agonist that induces vasoconstriction. It is commonly used in research related to nasal and conjunctival congestion.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07847