Learn More
Medchemexpress LLC Tetrahydrozoline hydrochloride | 522-48-5 | 99.9% | 1 G
Supplier: Medchemexpress LLC HYB0556A1G
Tetrahydrozoline hydrochloride, also known as Tetryzoline hydrochloride, is an imidazoline derivative. It acts as an α-adrenergic agonist, leading to vasoconstriction. This compound is frequently utilized in research studies focusing on nasal congestion and conjunctival congestion.
- Functions as an α-adrenergic agonist.
- Induces vasoconstriction.
- Used in research for nasal and conjunctival congestion.
- High purity (99.88%).
- Appears as a white to off-white solid.
- Exhibits good solubility in water and DMSO.
- Induces collagen types I and III synthesis in primary human gingival fibroblasts in vitro.