Learn More
Strem, An Ascensus Company CAS# 59091-96-2. 1g. cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II), 98%. MFCD00050955
SDP
Supplier:  Strem, An Ascensus Company 4470001G
Encompass
CAS# 59091-96-2. 1g. cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II), 98%. MFCD00050955. Molecular Weight: 484.49. Molecular Formula: RuCl2[CH3S(O)CH3]4. Color/form: yellow pwdr. Strem# 44-7000. http://www.strem.com/catalog/v/44-7000/
Catalog No. 50-901-12016