Learn More
Strem, An Ascensus Company CAS# 14221-01-3. Tetrakis(triphenylphosphine)palladium(0), 99% (99.9+%-Pd) MFCD00010012. 1G
SDP
Supplier:  Strem, An Ascensus Company 4621501G
Encompass
CAS# 14221-01-3. 1G. Tetrakis(triphenylphosphine)palladium(0), 99% (99.9+%-Pd). MFCD00010012. Molecular Formula: C72H60P4Pd. Molecular Weight: 1155.57. Color/Form: bright yellow pwdr. Stability: air sensitive, store cold (-18°C) Strem#: 46-2150 www.strem.com/catalog/v3/46-2150/
Catalog No. 50-901-12478