Learn More
Medchemexpress LLC Tetrakis(triphenylphosphine)palladium(0) | 14221-01-3 | MFCD00010012 | 5g
SDP
Supplier:  Medchemexpress LLC HYY17385G
Encompass_Preferred
Tetrakis(triphenylphosphine)palladium is a catalyst Tetrakis(triphenylphosphine)palladium catalyzes the highly regioselective addition of phenyl thiocyanate (PhSCN) to terminal alkynes[1]
Catalog No. 50-002-66578