Learn More
Medchemexpress LLC Tetrakis(triphenylphosphine)palladium | 14221-01-3 | MFCD00010012 | 98.0% | 10g
SDP
Supplier:  Medchemexpress LLC HYY173810G
Encompass_Preferred
Tetrakis(triphenylphosphine)palladium is a catalyst Tetrakis(triphenylphosphine)palladium catalyzes the highly regioselective addition of phenyl thiocyanate (PhSCN) to terminal alkynes[1]
Catalog No. 50-002-66166