Learn More
Medchemexpress LLC Tetramethylammonium chloride | 75-57-0 | MFCD00011628 | 98.0% | 109.60 | 50 G

Supplier: Medchemexpress LLC HYW012954B50G
TMA chloride (Tetramethylammonium chloride) is a chemical compound primarily used in molecular biology research. It is known to increase the specificity of hybridization and raise the DNA denaturation temperature during PCR processes. This compound is a white to off-white solid and is intended for research purposes only.
- Enhances hybridization specificity
- Elevates DNA denaturation temperature in PCR
- Inhibits TEA uptake in OCT1-expressing MDCK cells
- Inhibits TEA uptake in OCT2-expressing MDCK cells
- Inhibits MPP+ uptake in Xenopus laevis oocytes
- Inhibits TEA uptake in Xenopus laevis oocytes
- Soluble in H2O (≥ 100 mg/mL)
- Soluble in DMSO (5.83 mg/mL)
- Ships at room temperature
- Store at room temperature, dry and cool
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.