Learn More
Apexbio Technology LLC Tetramethylpyrazine 1124-11-4 20mg
SDP
Supplier:  Apexbio Technology LLC N191820
Encompass_Preferred
Tetramethylpyrazine (CAS 1124-11-4) a bioactive alkaloid primarily isolated from Ligusticum chuanxiong Hort exhibits multiple biological activities relevant to biomedical research Mechanistically it has been reported to modulate vascular smooth muscle contraction inhibit platelet aggregation and attenuate oxidative stress by scavenging reactive oxygen species These properties underlie its widespread application in studies of cardiovascular cerebrovascular and neurodegenerative diseases Tetramethylpyrazine can be solubilized in hot water or petroleum ether and should be stored in sealed cool and dry conditions to maintain stability during experimental use
Catalog No. 50-000-96533