Learn More
Selleck Chemical LLC Tetramethylpyrazine S3956-25mg
SDP
Supplier:  Selleck Chemical LLC S395625MG
Encompass_Preferred
Tetramethylpyrazine (ligustrazine TMP) is a natural compound isolated from Chinese herbal medicine Ligusticum wallichii (Chuan Xiong) with anti-inflammation antioxidant antiplatelet and antiapoptosis activities
Catalog No. 50-310-9153